Learn More
Chemglass Life Sciences O-Ring, Perfluoro (FFKM), #031, White - O-ring for laboratory glassware and fittings
SDP
Catalog No. 502743575
Encompass_Preferred
Perfluoro (FFKM) O-ring size #031 provides high chemical resistance and thermal stability for seals in laboratory glassware and fittings.
  • Size: #031.
  • Material: perfluoro (FFKM).
  • Color: white.
  • Durometer: 75 (shore).
  • Temperature range: -7 to 230° C.
  • Packaging: each.
  • Recommended compatibility: acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.
Catalog No. 50-274-3575 Supplier Chemglass Life Sciences Supplier No. CG309U031
Perfluoro (FFKM) O-ring size #031 provides high chemical resistance and thermal stability for seals in laboratory glassware and fittings.
  • Size: #031.
  • Material: perfluoro (FFKM).
  • Color: white.
  • Durometer: 75 (shore).
  • Temperature range: -7 to 230° C.
  • Packaging: each.
  • Recommended compatibility: acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.