Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #104, White
SDP
Catalog No. 502743573
Encompass_Preferred
Chemglass perfluoro (FFKM) O-ring, size #104, white, is a chemically resistant, thermally stable sealing component designed for laboratory and process equipment. It provides broad compatibility with aggressive solvents and corrosive media and is suitable for use in fittings, valves, and glassware assemblies.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • Effective service temperature: -7 to 230° C.
  • Compatible with acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Sold individually for easy replacement and inventory control.
Catalog No. 50-274-3573 Supplier Chemglass Life Sciences Supplier No. CG309U104
Chemglass perfluoro (FFKM) O-ring, size #104, white, is a chemically resistant, thermally stable sealing component designed for laboratory and process equipment. It provides broad compatibility with aggressive solvents and corrosive media and is suitable for use in fittings, valves, and glassware assemblies.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • Effective service temperature: -7 to 230° C.
  • Compatible with acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Sold individually for easy replacement and inventory control.