Learn More
Chemglass Life Sciences Perfluoro (FFKM) O-Ring, #105, White - For laboratory glassware seals
SDP
Catalog No. 502743572
Encompass_Preferred
Perfluoroelastomer (FFKM) O-ring designed for reliable sealing in aggressive chemical environments. This white O-ring is typically stocked at 75 durometer and provides wide temperature resistance for laboratory and process equipment.
  • Provides superior chemical resistance.
  • Wide operating temperature range (-7 to 230 °C).
  • Typical hardness of 75 durometer for reliable sealing.
  • White color for easy visual identification.
  • Suitable for sealing laboratory glassware joints and fittings.
Catalog No. 50-274-3572 Supplier Chemglass Life Sciences Supplier No. CG309U105
Perfluoroelastomer (FFKM) O-ring designed for reliable sealing in aggressive chemical environments. This white O-ring is typically stocked at 75 durometer and provides wide temperature resistance for laboratory and process equipment.
  • Provides superior chemical resistance.
  • Wide operating temperature range (-7 to 230 °C).
  • Typical hardness of 75 durometer for reliable sealing.
  • White color for easy visual identification.
  • Suitable for sealing laboratory glassware joints and fittings.