Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #107, White, 75 durometer
SDP
Catalog No. 502743571
Encompass_Preferred
Perfluoro (FFKM) O-ring for demanding chemical and high-temperature sealing applications. Stocked in white at 75 durometer and sold individually; effective service temperature -7 to 230 °C. Provides broad compatibility with acids, bases, alcohols, ethers, hydrocarbons, esters, aldehydes, ketones, amines, and chlorocarbon solvents.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • 75 durometer (white) for reliable sealing performance.
  • Effective service temperature -7 to 230 °C.
  • Suitable for acids, bases, alcohols, ethers, and a wide range of solvents.
  • Packaged and sold as individual units.
Catalog No. 50-274-3571 Supplier Chemglass Life Sciences Supplier No. CG309U107
Perfluoro (FFKM) O-ring for demanding chemical and high-temperature sealing applications. Stocked in white at 75 durometer and sold individually; effective service temperature -7 to 230 °C. Provides broad compatibility with acids, bases, alcohols, ethers, hydrocarbons, esters, aldehydes, ketones, amines, and chlorocarbon solvents.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • 75 durometer (white) for reliable sealing performance.
  • Effective service temperature -7 to 230 °C.
  • Suitable for acids, bases, alcohols, ethers, and a wide range of solvents.
  • Packaged and sold as individual units.