Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #108 - replacement seal for reaction vessels and lids
SDP
Catalog No. 502743570
Encompass_Preferred
O-Ring made from Perfluoro (FFKM) perfluoroelastomer for high chemical resistance and elevated temperature sealing applications. Typically used as a replacement seal in laboratory glassware, reaction vessels, lids, and fittings.
  • Perfluoro (FFKM) material provides superior chemical resistance.
  • Effective service temperature range of -7 to 230° C.
  • Typically stocked in white 75 durometer elastomer.
  • Sold individually (each).
  • Compatible with acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Suitable for replacement sealing in reaction vessels, lids, and laboratory fittings.
Catalog No. 50-274-3570 Supplier Chemglass Life Sciences Supplier No. CG309U108
O-Ring made from Perfluoro (FFKM) perfluoroelastomer for high chemical resistance and elevated temperature sealing applications. Typically used as a replacement seal in laboratory glassware, reaction vessels, lids, and fittings.
  • Perfluoro (FFKM) material provides superior chemical resistance.
  • Effective service temperature range of -7 to 230° C.
  • Typically stocked in white 75 durometer elastomer.
  • Sold individually (each).
  • Compatible with acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Suitable for replacement sealing in reaction vessels, lids, and laboratory fittings.