Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-ring, Size #109, White (CG-309-U-109)
SDP
Catalog No. 502743569
Encompass_Preferred
The Chemglass Perfluoro (FFKM) O-ring #109 is a high-performance perfluoroelastomer seal designed for demanding laboratory applications that require broad chemical compatibility and thermal stability. It is stocked in white, 75 durometer, and sold individually.
  • Made from perfluoroelastomer (FFKM).
  • White color with 75 durometer.
  • Effective temperature range -7 to 230 °C.
  • Resistant to acids, inorganic bases, alcohols, ethers, aromatic hydrocarbons, esters, aldehydes, ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Sold each (single unit).
Catalog No. 50-274-3569 Supplier Chemglass Life Sciences Supplier No. CG309U109
The Chemglass Perfluoro (FFKM) O-ring #109 is a high-performance perfluoroelastomer seal designed for demanding laboratory applications that require broad chemical compatibility and thermal stability. It is stocked in white, 75 durometer, and sold individually.
  • Made from perfluoroelastomer (FFKM).
  • White color with 75 durometer.
  • Effective temperature range -7 to 230 °C.
  • Resistant to acids, inorganic bases, alcohols, ethers, aromatic hydrocarbons, esters, aldehydes, ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Sold each (single unit).