Learn More
Chemglass Life Sciences O-Ring, Perfluoro (FFKM), #111, White
SDP
Catalog No. 502743568
Encompass_Preferred
Chemglass perfluoro (FFKM) O-ring, size #111 in white, provides high chemical resistance and thermal stability for demanding sealing applications. Manufactured from perfluoroelastomer at 75 durometer, it is suitable for service temperatures from -7 to 230 °C and compatible with a wide range of solvents and corrosive media.
  • Perfluoro (FFKM) material for broad chemical resistance.
  • White, 75 durometer construction.
  • Service temperature -7 to 230 °C.
  • Available in AS568 size #111.
  • Suitable for acids, bases, solvents, and amines.
Catalog No. 50-274-3568 Supplier Chemglass Life Sciences Supplier No. CG309U111
Chemglass perfluoro (FFKM) O-ring, size #111 in white, provides high chemical resistance and thermal stability for demanding sealing applications. Manufactured from perfluoroelastomer at 75 durometer, it is suitable for service temperatures from -7 to 230 °C and compatible with a wide range of solvents and corrosive media.
  • Perfluoro (FFKM) material for broad chemical resistance.
  • White, 75 durometer construction.
  • Service temperature -7 to 230 °C.
  • Available in AS568 size #111.
  • Suitable for acids, bases, solvents, and amines.