Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-Ring, Size 114, White, 75 Durometer
SDP
Catalog No. 502743566
Encompass_Preferred
Perfluoro (FFKM) O-ring engineered for high chemical resistance and thermal stability. Intended for sealing applications in aggressive chemical environments, laboratory glassware, and process equipment where durability and long service life are required.
  • Perfluoro (FFKM) elastomer material for broad chemical compatibility.
  • White 75 durometer compound for high thermal stability.
  • Effective service temperature -7 to 230° C.
  • Recommended for acids, inorganic bases, alcohols/ethers, aromatic and aliphatic hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents.
  • Suitable for sealing glassware joints, flanges, and general laboratory fittings.
Catalog No. 50-274-3566 Supplier Chemglass Life Sciences Supplier No. CG309U114
Perfluoro (FFKM) O-ring engineered for high chemical resistance and thermal stability. Intended for sealing applications in aggressive chemical environments, laboratory glassware, and process equipment where durability and long service life are required.
  • Perfluoro (FFKM) elastomer material for broad chemical compatibility.
  • White 75 durometer compound for high thermal stability.
  • Effective service temperature -7 to 230° C.
  • Recommended for acids, inorganic bases, alcohols/ethers, aromatic and aliphatic hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents.
  • Suitable for sealing glassware joints, flanges, and general laboratory fittings.