Learn More
Chemglass Life Sciences O-Ring, Perfluoro (FFKM), #117, White - Seal for reaction vessels and lids
SDP
Catalog No. 502743565
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #117, provides exceptional chemical and thermal resistance for reliable sealing in laboratory glassware, reaction vessels, and process equipment. The white FFKM compound maintains elasticity across a wide temperature range and is suitable for aggressive chemical environments.
  • Provides superior chemical resistance to acids, bases, alcohols, ethers, aromatic and aliphatic hydrocarbons, esters, ketones, and chlorinated solvents.
  • Effective service temperature range of -7 to 230 °C.
  • White perfluoroelastomer construction with 75 durometer for resilient sealing.
  • Suitable for sealing reaction vessels, lids, and other laboratory glassware.
  • Supplied individually (each) for convenient replacement.
Catalog No. 50-274-3565 Supplier Chemglass Life Sciences Supplier No. CG309U117
Perfluoro (FFKM) O-ring, size #117, provides exceptional chemical and thermal resistance for reliable sealing in laboratory glassware, reaction vessels, and process equipment. The white FFKM compound maintains elasticity across a wide temperature range and is suitable for aggressive chemical environments.
  • Provides superior chemical resistance to acids, bases, alcohols, ethers, aromatic and aliphatic hydrocarbons, esters, ketones, and chlorinated solvents.
  • Effective service temperature range of -7 to 230 °C.
  • White perfluoroelastomer construction with 75 durometer for resilient sealing.
  • Suitable for sealing reaction vessels, lids, and other laboratory glassware.
  • Supplied individually (each) for convenient replacement.