Learn More
Chemglass Life Sciences Chemglass O-ring, Perfluoro (FFKM), #119, White
SDP
Catalog No. 502743564
Encompass_Preferred
Perfluoro (FFKM) o-ring designed for chemical-resistant sealing in laboratory equipment. The material offers high thermal stability and broad chemical compatibility, suitable for applications requiring resistance to aggressive solvents and high temperatures.
  • Material: perfluoro (FFKM).
  • Color: white.
  • Size/number: #119.
  • Durometer: 75 (white stock).
  • Temperature range: -7 to 230° C.
  • Recommended media: acids; inorganic bases; alcohols and ethers; aromatic hydrocarbons; esters; aldehydes and ketones; aliphatic hydrocarbons; amines; chlorocarbon solvents.
  • Dimensions: not listed on manufacturer page.
Catalog No. 50-274-3564 Supplier Chemglass Life Sciences Supplier No. CG309U119
Perfluoro (FFKM) o-ring designed for chemical-resistant sealing in laboratory equipment. The material offers high thermal stability and broad chemical compatibility, suitable for applications requiring resistance to aggressive solvents and high temperatures.
  • Material: perfluoro (FFKM).
  • Color: white.
  • Size/number: #119.
  • Durometer: 75 (white stock).
  • Temperature range: -7 to 230° C.
  • Recommended media: acids; inorganic bases; alcohols and ethers; aromatic hydrocarbons; esters; aldehydes and ketones; aliphatic hydrocarbons; amines; chlorocarbon solvents.
  • Dimensions: not listed on manufacturer page.