Learn More
Chemglass Life Sciences O-Ring, Perfluoro (FFKM), #120, White - replacement O-ring for Chemglass fittings and glassware
SDP
Catalog No. 502743563
Encompass_Preferred
Perfluoro (FFKM) O-ring designed for chemical- and temperature-resistant sealing in laboratory glassware and fittings. Suitable for use where aggressive chemicals or elevated temperatures are present.
  • Perfluoroelastomer (FFKM) material provides superior chemical resistance.
  • White color for visual identification.
  • Operational temperature range of -7 to 230° C.
  • Sold individually (each).
  • Suitable for sealing glassware joints, fittings, and vacuum applications.
Catalog No. 50-274-3563 Supplier Chemglass Life Sciences Supplier No. CG309U120
Perfluoro (FFKM) O-ring designed for chemical- and temperature-resistant sealing in laboratory glassware and fittings. Suitable for use where aggressive chemicals or elevated temperatures are present.
  • Perfluoroelastomer (FFKM) material provides superior chemical resistance.
  • White color for visual identification.
  • Operational temperature range of -7 to 230° C.
  • Sold individually (each).
  • Suitable for sealing glassware joints, fittings, and vacuum applications.