Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), Size 121, White, 75 durometer
SDP
Catalog No. 502743562
Encompass_Preferred
Chemglass perfluoro (FFKM) O-ring, size 121, is a high-performance sealing element engineered for aggressive chemical and elevated temperature environments. It is produced in a white, high-purity FFKM compound and provides broad chemical resistance and thermal stability.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • Suitable for high temperature applications: -7 to 230° C.
  • Stocked white, 75 durometer for general sealing needs.
  • Intended for use as a sealing O-ring in laboratory and process equipment.
Catalog No. 50-274-3562 Supplier Chemglass Life Sciences Supplier No. CG309U121
Chemglass perfluoro (FFKM) O-ring, size 121, is a high-performance sealing element engineered for aggressive chemical and elevated temperature environments. It is produced in a white, high-purity FFKM compound and provides broad chemical resistance and thermal stability.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • Suitable for high temperature applications: -7 to 230° C.
  • Stocked white, 75 durometer for general sealing needs.
  • Intended for use as a sealing O-ring in laboratory and process equipment.