Learn More
Chemglass Life Sciences Perfluoro (FFKM) O-Ring, Size 122 - White; For Laboratory Reaction Vessels and Lids
SDP
Catalog No. 502743561
Encompass_Preferred
This Chemglass perfluoro (FFKM) O-ring, size number 122, provides high chemical and thermal resistance for seals used in laboratory reaction vessels, lids, and glassware. It is supplied in white 75 durometer material and is intended for aggressive chemical environments and elevated temperatures.
  • Made from perfluoro (FFKM) elastomer.
  • White color, 75 durometer.
  • Service temperature -7-230 °C.
  • High resistance to acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Suitable as a replacement seal for reaction vessels, lids, and laboratory glassware.
Catalog No. 50-274-3561 Supplier Chemglass Life Sciences Supplier No. CG309U122
This Chemglass perfluoro (FFKM) O-ring, size number 122, provides high chemical and thermal resistance for seals used in laboratory reaction vessels, lids, and glassware. It is supplied in white 75 durometer material and is intended for aggressive chemical environments and elevated temperatures.
  • Made from perfluoro (FFKM) elastomer.
  • White color, 75 durometer.
  • Service temperature -7-230 °C.
  • High resistance to acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Suitable as a replacement seal for reaction vessels, lids, and laboratory glassware.