Learn More
Chemglass Life Sciences Chemglass perfluoro (FFKM) O-ring, size #129, 75 durometer - replacement for reaction vessels and laboratory glassware
SDP
Catalog No. 502743559
Encompass_Preferred
Chemglass perfluoroelastomer (FFKM) O-ring, size #129, provides high thermal stability and broad chemical resistance for demanding laboratory sealing applications. It is intended as a replacement O-ring for reaction vessels, lids, and other glassware exposed to aggressive solvents or elevated temperatures.
  • Perfluoro (FFKM) material for broad chemical resistance.
  • 75 durometer hardness for durable sealing performance.
  • White color for easy visual identification.
  • Compatible with acids, inorganic bases, alcohols, ethers, aromatic hydrocarbons, esters, aldehydes, ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Sized as a #129 O-ring for replacement use in compatible assemblies.
Catalog No. 50-274-3559 Supplier Chemglass Life Sciences Supplier No. CG309U129
Chemglass perfluoroelastomer (FFKM) O-ring, size #129, provides high thermal stability and broad chemical resistance for demanding laboratory sealing applications. It is intended as a replacement O-ring for reaction vessels, lids, and other glassware exposed to aggressive solvents or elevated temperatures.
  • Perfluoro (FFKM) material for broad chemical resistance.
  • 75 durometer hardness for durable sealing performance.
  • White color for easy visual identification.
  • Compatible with acids, inorganic bases, alcohols, ethers, aromatic hydrocarbons, esters, aldehydes, ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Sized as a #129 O-ring for replacement use in compatible assemblies.