Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-Ring, Size #132, White
SDP
Catalog No. 502743558
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #132, is a perfluoroelastomer sealing ring designed for high chemical and thermal resistance in laboratory glassware, fittings, and process equipment.
  • Material: FFKM (perfluoroelastomer).
  • Size: #132.
  • Color: White.
  • Durometer: 75 (stocked).
  • Effective service temperature: -7 to 230 °C.
  • Chemical compatibility: acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.
  • Packaging: sold each; call for availability.
Catalog No. 50-274-3558 Supplier Chemglass Life Sciences Supplier No. CG309U132
Perfluoro (FFKM) O-ring, size #132, is a perfluoroelastomer sealing ring designed for high chemical and thermal resistance in laboratory glassware, fittings, and process equipment.
  • Material: FFKM (perfluoroelastomer).
  • Size: #132.
  • Color: White.
  • Durometer: 75 (stocked).
  • Effective service temperature: -7 to 230 °C.
  • Chemical compatibility: acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.
  • Packaging: sold each; call for availability.