Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), Size #136, White - Replacement sealing O-ring for laboratory glassware and fittings
SDP
Catalog No. 502743557
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #136, in white. Made from FFKM perfluoroelastomer, this O-ring offers superior chemical and thermal resistance for demanding laboratory sealing applications.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance.
  • Effective service temperature range: -7 to 230 °C.
  • White 75 durometer for consistent sealing performance.
  • Suitable for acids, inorganic bases, alcohols/ethers, hydrocarbons, esters, ketones, amines, and chlorocarbon solvents.
  • Intended as a replacement sealing O-ring for laboratory glassware and fittings.
Catalog No. 50-274-3557 Supplier Chemglass Life Sciences Supplier No. CG309U136
Perfluoro (FFKM) O-ring, size #136, in white. Made from FFKM perfluoroelastomer, this O-ring offers superior chemical and thermal resistance for demanding laboratory sealing applications.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance.
  • Effective service temperature range: -7 to 230 °C.
  • White 75 durometer for consistent sealing performance.
  • Suitable for acids, inorganic bases, alcohols/ethers, hydrocarbons, esters, ketones, amines, and chlorocarbon solvents.
  • Intended as a replacement sealing O-ring for laboratory glassware and fittings.