Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoroelastomer (FFKM), size #137, white
SDP
Catalog No. 502743556
Encompass_Preferred
Chemglass CG-309-U-137 is a perfluoro (FFKM) O-ring in white designed for demanding chemical sealing applications. It delivers broad chemical compatibility and high-temperature resistance for use in laboratory glassware, valves, and fittings.
  • Perfluoroelastomer (FFKM) construction for broad chemical compatibility.
  • White, 75 durometer for durable sealing performance.
  • Wide service temperature range suitable for varied process conditions.
  • Resistant to acids, bases, solvents, and hydrocarbons.
  • Used as a replacement seal in valves, fittings, and glass assemblies.
Catalog No. 50-274-3556 Supplier Chemglass Life Sciences Supplier No. CG309U137
Chemglass CG-309-U-137 is a perfluoro (FFKM) O-ring in white designed for demanding chemical sealing applications. It delivers broad chemical compatibility and high-temperature resistance for use in laboratory glassware, valves, and fittings.
  • Perfluoroelastomer (FFKM) construction for broad chemical compatibility.
  • White, 75 durometer for durable sealing performance.
  • Wide service temperature range suitable for varied process conditions.
  • Resistant to acids, bases, solvents, and hydrocarbons.
  • Used as a replacement seal in valves, fittings, and glass assemblies.