Learn More
Chemglass Life Sciences O-Ring, Perfluoro (FFKM), #201, White
SDP
Catalog No. 502743555
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #201, white. The perfluoroelastomer compound provides high thermal stability and broad chemical resistance for sealing applications in laboratory glassware and equipment.
  • Perfluoroelastomer material for high chemical resistance.
  • Standard size O-ring for common laboratory fittings.
  • White color for visual identification.
  • 75 durometer hardness.
  • Service temperature -7 to 230 °C.
  • Compatible with acids, inorganic bases, alcohols and ethers, aromatic hydrocarbons, esters, aldehydes and ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Sold individually.
Catalog No. 50-274-3555 Supplier Chemglass Life Sciences Supplier No. CG309U201
Perfluoro (FFKM) O-ring, size #201, white. The perfluoroelastomer compound provides high thermal stability and broad chemical resistance for sealing applications in laboratory glassware and equipment.
  • Perfluoroelastomer material for high chemical resistance.
  • Standard size O-ring for common laboratory fittings.
  • White color for visual identification.
  • 75 durometer hardness.
  • Service temperature -7 to 230 °C.
  • Compatible with acids, inorganic bases, alcohols and ethers, aromatic hydrocarbons, esters, aldehydes and ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Sold individually.