Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #204, White
SDP
Catalog No. 502743554
Encompass_Preferred
Chemglass Perfluoro (FFKM) O-ring, size #204 (white), offers high thermal stability and broad chemical resistance for laboratory sealing applications where standard elastomers fail.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • White color for visual identification.
  • Size #204, standard o-ring dimension.
  • Service temperature -7 to 230° C.
  • Recommended for acids, inorganic bases, alcohols and ethers, aromatic hydrocarbons, esters, aldehydes and ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
Catalog No. 50-274-3554 Supplier Chemglass Life Sciences Supplier No. CG309U204
Chemglass Perfluoro (FFKM) O-ring, size #204 (white), offers high thermal stability and broad chemical resistance for laboratory sealing applications where standard elastomers fail.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • White color for visual identification.
  • Size #204, standard o-ring dimension.
  • Service temperature -7 to 230° C.
  • Recommended for acids, inorganic bases, alcohols and ethers, aromatic hydrocarbons, esters, aldehydes and ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.