Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #205, White - seals laboratory glassware joints and fittings.
SDP
Catalog No. 502743553
Encompass_Preferred
Perfluoro (FFKM) O-ring engineered for demanding chemical environments; provides broad chemical resistance and elevated temperature performance for sealing laboratory glassware, fittings, and process equipment.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • White color.
  • Size number: 205.
  • 75 durometer.
  • Effective service temperature: -7 to 230 °C.
  • Suitable for acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Call for availability.
Catalog No. 50-274-3553 Supplier Chemglass Life Sciences Supplier No. CG309U205
Perfluoro (FFKM) O-ring engineered for demanding chemical environments; provides broad chemical resistance and elevated temperature performance for sealing laboratory glassware, fittings, and process equipment.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • White color.
  • Size number: 205.
  • 75 durometer.
  • Effective service temperature: -7 to 230 °C.
  • Suitable for acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Call for availability.