Learn More
Chemglass Life Sciences Perfluoro (FFKM) O-ring, size #212, white - replacement seal for laboratory glassware and fittings
SDP
Catalog No. 502743551
Encompass_Preferred
The Chemglass perfluoro (FFKM) O-ring, size #212, is a white perfluoroelastomer seal designed for demanding laboratory applications that require exceptional chemical and thermal resistance. It is intended as a replacement seal for laboratory glassware and fittings where compatibility with aggressive solvents and high temperatures is required.
  • High chemical resistance suitable for acids, bases, and a wide range of solvents.
  • Excellent thermal stability for high-temperature laboratory applications.
  • White color for quick visual identification and contamination control.
  • Perfluoroelastomer (FFKM) construction provides long service life in aggressive environments.
  • Suitable for use as a replacement seal in laboratory glassware and fittings.
Catalog No. 50-274-3551 Supplier Chemglass Life Sciences Supplier No. CG309U212
The Chemglass perfluoro (FFKM) O-ring, size #212, is a white perfluoroelastomer seal designed for demanding laboratory applications that require exceptional chemical and thermal resistance. It is intended as a replacement seal for laboratory glassware and fittings where compatibility with aggressive solvents and high temperatures is required.
  • High chemical resistance suitable for acids, bases, and a wide range of solvents.
  • Excellent thermal stability for high-temperature laboratory applications.
  • White color for quick visual identification and contamination control.
  • Perfluoroelastomer (FFKM) construction provides long service life in aggressive environments.
  • Suitable for use as a replacement seal in laboratory glassware and fittings.