Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), White, Size 214 - CG-309-U-214
SDP
Catalog No. 502743550
Encompass_Preferred
Perfluoro (FFKM) O-ring for high-performance sealing applications; white, size 214, provides high thermal stability and broad chemical resistance suitable for laboratory and process use.
  • Provides superior chemical resistance to acids, bases, and common solvents.
  • Wide service temperature range from -7 to 230° C.
  • White perfluoro (FFKM) formulation for high-purity applications.
  • Compatible with aggressive chemistries including chlorinated solvents and amines.
  • Suitable for seals and fittings in laboratory and process equipment.
Catalog No. 50-274-3550 Supplier Chemglass Life Sciences Supplier No. CG309U214
Perfluoro (FFKM) O-ring for high-performance sealing applications; white, size 214, provides high thermal stability and broad chemical resistance suitable for laboratory and process use.
  • Provides superior chemical resistance to acids, bases, and common solvents.
  • Wide service temperature range from -7 to 230° C.
  • White perfluoro (FFKM) formulation for high-purity applications.
  • Compatible with aggressive chemistries including chlorinated solvents and amines.
  • Suitable for seals and fittings in laboratory and process equipment.