Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #218, White (CG-309-U-218)
SDP
Catalog No. 502743548
Encompass_Preferred
Chemglass Life Sciences perfluoro (FFKM) O-ring, size #218, is a thermally stable sealing component suitable for laboratory and chemical-processing applications. Perfluoro (FFKM) offers superior chemical resistance and a broad operating temperature range compared with many other elastomers.
  • Material: perfluoro (FFKM).
  • Color: white.
  • Size/number: #218.
  • Durometer: 75 (white standard).
  • Temperature range: -7 to 230 °C.
  • Recommended for use with acids, inorganic bases, alcohols/ethers, aromatic and aliphatic hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents.
  • Packaging: each.
Catalog No. 50-274-3548 Supplier Chemglass Life Sciences Supplier No. CG309U218
Chemglass Life Sciences perfluoro (FFKM) O-ring, size #218, is a thermally stable sealing component suitable for laboratory and chemical-processing applications. Perfluoro (FFKM) offers superior chemical resistance and a broad operating temperature range compared with many other elastomers.
  • Material: perfluoro (FFKM).
  • Color: white.
  • Size/number: #218.
  • Durometer: 75 (white standard).
  • Temperature range: -7 to 230 °C.
  • Recommended for use with acids, inorganic bases, alcohols/ethers, aromatic and aliphatic hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents.
  • Packaging: each.