Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences O-Ring, Perfluoro (FFKM), Size 219 - Replacement sealing ring for Chemglass fittings
SDP

Catalog No. 502743547
Encompass_Preferred

O-Ring perfluoro (FFKM) size 219 is a thermally stable perfluoroelastomer sealing ring designed for demanding laboratory and process applications that require broad chemical compatibility and high temperature resistance.

  • Material: perfluoroelastomer (FFKM)
  • Size number: 219
  • Color: white
  • Durometer: 75 durometer
  • Service temperature: -7 to 230° C
  • Applications: acids, inorganic bases, alcohols and ethers, aromatic hydrocarbons, esters, aldehydes and ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents

Catalog No. 50-274-3547 Supplier Chemglass Life Sciences Supplier No. CG309U219
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-Ring perfluoro (FFKM) size 219 is a thermally stable perfluoroelastomer sealing ring designed for demanding laboratory and process applications that require broad chemical compatibility and high temperature resistance.

  • Material: perfluoroelastomer (FFKM)
  • Size number: 219
  • Color: white
  • Durometer: 75 durometer
  • Service temperature: -7 to 230° C
  • Applications: acids, inorganic bases, alcohols and ethers, aromatic hydrocarbons, esters, aldehydes and ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.