Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-Ring, Size #222, White - Replacement O-ring for laboratory fittings
SDP
Catalog No. 502743545
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #222, white, engineered for broad chemical resistance and elevated temperature service in laboratory and chemical-processing seals.
  • Material: perfluoro (FFKM).
  • Color: white.
  • Hardness: 75 durometer (stocked).
  • Service temperature: -7 to 230 °C.
  • Chemical resistance: suitable for acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Typical applications: seals for glassware joints, valves, and chemical processing equipment.
Catalog No. 50-274-3545 Supplier Chemglass Life Sciences Supplier No. CG309U222
Perfluoro (FFKM) O-ring, size #222, white, engineered for broad chemical resistance and elevated temperature service in laboratory and chemical-processing seals.
  • Material: perfluoro (FFKM).
  • Color: white.
  • Hardness: 75 durometer (stocked).
  • Service temperature: -7 to 230 °C.
  • Chemical resistance: suitable for acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Typical applications: seals for glassware joints, valves, and chemical processing equipment.