Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), White, size #224
SDP
Catalog No. 502743543
Encompass_Preferred
Chemglass Perfluoro (FFKM) O-ring #224 (CG-309-U-224) is a white, 75 durometer perfluoroelastomer O-ring designed for demanding chemical and thermal service. It provides broad chemical compatibility and thermal stability for laboratory glassware, valves, and fluid transfer applications where aggressive media are present.
  • Perfluoro (FFKM) construction for superior chemical resistance.
  • White color with 75 durometer hardness.
  • Effective service temperature: -7 to 230° C.
  • Suitable for acids, inorganic bases, alcohols, ethers, aromatic and aliphatic hydrocarbons, esters, aldehydes, ketones, amines, and chlorocarbon solvents.
  • Stocked in dash size #224 for direct replacement use.
Catalog No. 50-274-3543 Supplier Chemglass Life Sciences Supplier No. CG309U224
Chemglass Perfluoro (FFKM) O-ring #224 (CG-309-U-224) is a white, 75 durometer perfluoroelastomer O-ring designed for demanding chemical and thermal service. It provides broad chemical compatibility and thermal stability for laboratory glassware, valves, and fluid transfer applications where aggressive media are present.
  • Perfluoro (FFKM) construction for superior chemical resistance.
  • White color with 75 durometer hardness.
  • Effective service temperature: -7 to 230° C.
  • Suitable for acids, inorganic bases, alcohols, ethers, aromatic and aliphatic hydrocarbons, esters, aldehydes, ketones, amines, and chlorocarbon solvents.
  • Stocked in dash size #224 for direct replacement use.