Learn More
Chemglass Life Sciences Perfluoro (FFKM) O-Ring, #225, White, 75 Durometer
SDP
Catalog No. 502743542
Encompass_Preferred
Perfluoro (FFKM) O-ring #225, white, is a chemically resistant sealing component engineered for demanding applications. It provides broad chemical compatibility and thermal stability for use in aggressive chemical environments.
  • Material: Perfluoro (FFKM).
  • Color: White.
  • Durometer: 75.
  • Effective service temperature range: -7 to 230° C.
  • Recommended for use with: acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.
  • Size reference: #225 o-ring size index.
Catalog No. 50-274-3542 Supplier Chemglass Life Sciences Supplier No. CG309U225
Perfluoro (FFKM) O-ring #225, white, is a chemically resistant sealing component engineered for demanding applications. It provides broad chemical compatibility and thermal stability for use in aggressive chemical environments.
  • Material: Perfluoro (FFKM).
  • Color: White.
  • Durometer: 75.
  • Effective service temperature range: -7 to 230° C.
  • Recommended for use with: acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.
  • Size reference: #225 o-ring size index.