Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-ring, size 226 (white, 75 durometer)
SDP

Catalog No. 502743541
Encompass_Preferred

Perfluoro (FFKM) O-ring, size 226, designed for demanding laboratory sealing applications. The perfluoroelastomer material provides exceptional chemical resistance and thermal stability for use with aggressive solvents and a wide range of reagents. Suitable for sealing glassware, valves, fittings, and reactors.

  • Material: perfluoroelastomer (FFKM).
  • Size: #226.
  • Color: white.
  • Durometer: 75 (typical).
  • Service temperature: -7 to 230 °C.
  • Chemical compatibility: acids, inorganic bases, alcohols/ethers, aromatic and aliphatic hydrocarbons, esters, aldehydes/ketones, amines, chlorinated solvents.
  • Applications: seals for glassware, valves, fittings, and reactors.

Catalog No. 50-274-3541 Supplier Chemglass Life Sciences Supplier No. CG309U226
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Perfluoro (FFKM) O-ring, size 226, designed for demanding laboratory sealing applications. The perfluoroelastomer material provides exceptional chemical resistance and thermal stability for use with aggressive solvents and a wide range of reagents. Suitable for sealing glassware, valves, fittings, and reactors.

  • Material: perfluoroelastomer (FFKM).
  • Size: #226.
  • Color: white.
  • Durometer: 75 (typical).
  • Service temperature: -7 to 230 °C.
  • Chemical compatibility: acids, inorganic bases, alcohols/ethers, aromatic and aliphatic hydrocarbons, esters, aldehydes/ketones, amines, chlorinated solvents.
  • Applications: seals for glassware, valves, fittings, and reactors.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.