Learn More
Chemglass Life Sciences Perfluoro (FFKM) O-Ring, AS568 #229, White (CG-309-U-229)
SDP
Catalog No. 502743540
Encompass_Preferred
Perfluoro (FFKM) O-ring in AS568 size #229 designed for demanding chemical and thermal applications. The perfluoroelastomer compound provides high resistance to a broad range of aggressive chemicals while maintaining serviceability across a wide temperature range.
  • Perfluoroelastomer (FFKM) material for superior chemical resistance.
  • White color for easy visual identification.
  • Service temperature range: -7 to 230° C.
  • Stocked at 75 durometer for standard sealing performance.
  • Suitable for acids; inorganic bases; alcohols and ethers; aromatic and aliphatic hydrocarbons; esters; aldehydes and ketones; amines; chlorocarbon solvents.
  • Suitable for replacement sealing in laboratory and process equipment.
Catalog No. 50-274-3540 Supplier Chemglass Life Sciences Supplier No. CG309U229
Perfluoro (FFKM) O-ring in AS568 size #229 designed for demanding chemical and thermal applications. The perfluoroelastomer compound provides high resistance to a broad range of aggressive chemicals while maintaining serviceability across a wide temperature range.
  • Perfluoroelastomer (FFKM) material for superior chemical resistance.
  • White color for easy visual identification.
  • Service temperature range: -7 to 230° C.
  • Stocked at 75 durometer for standard sealing performance.
  • Suitable for acids; inorganic bases; alcohols and ethers; aromatic and aliphatic hydrocarbons; esters; aldehydes and ketones; amines; chlorocarbon solvents.
  • Suitable for replacement sealing in laboratory and process equipment.