Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #326, White
SDP
Catalog No. 502743539
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #326, white, designed for demanding chemical and high-temperature sealing applications. The perfluoroelastomer construction provides superior thermal stability and broad resistance to aggressive solvents and reagents.
  • Perfluoro (FFKM) material for broad chemical resistance.
  • White color with 75 durometer hardness.
  • Effective service temperature -7 to 230 °C.
  • Suitable for acids, inorganic bases, alcohols, ethers, ketones, aromatic and aliphatic hydrocarbons, amines, and chlorinated solvents.
  • Stocked and sold individually for replacement and maintenance.
Catalog No. 50-274-3539 Supplier Chemglass Life Sciences Supplier No. CG309U326
Perfluoro (FFKM) O-ring, size #326, white, designed for demanding chemical and high-temperature sealing applications. The perfluoroelastomer construction provides superior thermal stability and broad resistance to aggressive solvents and reagents.
  • Perfluoro (FFKM) material for broad chemical resistance.
  • White color with 75 durometer hardness.
  • Effective service temperature -7 to 230 °C.
  • Suitable for acids, inorganic bases, alcohols, ethers, ketones, aromatic and aliphatic hydrocarbons, amines, and chlorinated solvents.
  • Stocked and sold individually for replacement and maintenance.