Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-Ring, Size #331, White - chemical- and heat-resistant seal for laboratory glassware joints
SDP
Catalog No. 502743538
Encompass_Preferred
Perfluoro (FFKM) o-ring, white, size #331, engineered for high chemical and thermal resistance in laboratory glassware seals and o-ring joints. Suitable for demanding chemical environments where broad solvent compatibility and elevated service temperature are required.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • White color for clear identification.
  • Effective service temperature range: -7 to 230° C.
  • Compatible with acids, inorganic bases, alcohols/ethers, hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents.
  • Designed for o-ring joints and seals in laboratory glassware and reaction vessels.
Catalog No. 50-274-3538 Supplier Chemglass Life Sciences Supplier No. CG309U331
Perfluoro (FFKM) o-ring, white, size #331, engineered for high chemical and thermal resistance in laboratory glassware seals and o-ring joints. Suitable for demanding chemical environments where broad solvent compatibility and elevated service temperature are required.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • White color for clear identification.
  • Effective service temperature range: -7 to 230° C.
  • Compatible with acids, inorganic bases, alcohols/ethers, hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents.
  • Designed for o-ring joints and seals in laboratory glassware and reaction vessels.