Learn More
Chemglass Life Sciences O-Ring, Perfluoro, #108
SDP
Catalog No. 50872245
Encompass_Preferred
O-Ring, Perfluoro, #108. One of the most thermally stable elastomeric materials available, Perfluoro is a family of perfluoroelastomer compounds designed to provide superior chemical resistance, and they offer a cost savings over similar o-rings such as Kalrez 4079 and Chemraz. Perfluoro o-rings are stocked in WHITE 75 durometer (unless otherwise noted). The effective service temperature is -7 to 230 C. Recommended for use with acids, inorganic bases, alcohols/ethers, aromatic hydro-carbons, esters, aldehydes/ ketones, aliphatic hydrocarbons, amines and chlorocarbon solvents. NOTE: FOR PERFLUORO O-RINGS USED WITH DURAN FLANGES, SEE CG-147.
Catalog No. 50-872-245 Supplier Chemglass Life Sciences Supplier No. CG309108
O-Ring, Perfluoro, #108. One of the most thermally stable elastomeric materials available, Perfluoro is a family of perfluoroelastomer compounds designed to provide superior chemical resistance, and they offer a cost savings over similar o-rings such as Kalrez 4079 and Chemraz. Perfluoro o-rings are stocked in WHITE 75 durometer (unless otherwise noted). The effective service temperature is -7 to 230 C. Recommended for use with acids, inorganic bases, alcohols/ethers, aromatic hydro-carbons, esters, aldehydes/ ketones, aliphatic hydrocarbons, amines and chlorocarbon solvents. NOTE: FOR PERFLUORO O-RINGS USED WITH DURAN FLANGES, SEE CG-147.