Learn More
Perkin Elmer US LLC O-Ring for Needle Seal Assembly, 10/pk
Catalog No. 509051999
Encompass_Preferred
TurboMatrix Headspace and high-sensitivity Headspace Trap samplers provide unparalleled precision and ease of use for numerous GC or GC/MS volatile-analysis applications. The system can manage up to 12 samples simultaneously, ensuring that the next sample is ready for analysis upon completion of the previous run, achieving significant time savings.
Catalog No. 50-905-1999 Supplier Perkin Elmer US LLC Supplier No. B0198110
TurboMatrix Headspace and high-sensitivity Headspace Trap samplers provide unparalleled precision and ease of use for numerous GC or GC/MS volatile-analysis applications. The system can manage up to 12 samples simultaneously, ensuring that the next sample is ready for analysis upon completion of the previous run, achieving significant time savings.