Learn More
Bellco Glass, Inc. O Ring, Platinum Cured Sillicone, 53mm, 5/PK
SDP
Supplier:  Bellco Glass, Inc. 452800427
Encompass_Preferred
Silicone Replacement O-Rings are constructed from USP Class VI Platinum-Cured Silicone. Intended for use with VersaCaps, these O-Rings ensure an excellent seal on a variety of containers, including Bottles, Flasks, and Carboys. The 53B O-Ring size can be used with 53B VersaCaps and VersaCap Adapters. For increased chemical compatibility, EPDM O-Rings are also available.
Catalog No. 50-255-0335