Learn More
Waters Corp Waters Corporation O-Ring Removal Tool, Brass, 1 per package
Catalog No. 50466625 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The O-Ring Removal Tool is a durable brass tool specifically designed for the efficient and safe removal of O-rings from various mass spectrometry (MS) systems. It is supplied as a single unit per package.
  • Constructed from durable brass.
  • Packaged as one unit.
  • Designed for use with mass spectrometry (MS) systems.
  • Compatible with numerous Waters MS instruments, including the 3100 Mass Detector, ACQUITY SQD System, Xevo TQ Absolute, BioAccord System, and SYNAPT series.
  • Requires replacement on an as-needed basis.
Catalog No. 50-466-625 Supplier Waters Corp Supplier No. 700005054
The O-Ring Removal Tool is a durable brass tool specifically designed for the efficient and safe removal of O-rings from various mass spectrometry (MS) systems. It is supplied as a single unit per package.
  • Constructed from durable brass.
  • Packaged as one unit.
  • Designed for use with mass spectrometry (MS) systems.
  • Compatible with numerous Waters MS instruments, including the 3100 Mass Detector, ACQUITY SQD System, Xevo TQ Absolute, BioAccord System, and SYNAPT series.
  • Requires replacement on an as-needed basis.