Learn More
Glass Expansion O-ring Removal tool
SDP
Catalog No. 502703097
Encompass_Preferred
O-ring Removal Tool. Facilitates the removal and installation of O-rings used with Glass Expansion D-Torches. This tool is especially handy while wearing PPE that can limit dexterity.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3097 Supplier Glass Expansion Supplier No. 501001636
O-ring Removal Tool. Facilitates the removal and installation of O-rings used with Glass Expansion D-Torches. This tool is especially handy while wearing PPE that can limit dexterity.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001