Learn More
Beckman Coulter Replacement O-Ring 111mm ID x 177.5mm OD
Catalog No. NC0111689
Encompass
Replacement O-ring for the centrifuge bucket cover assembly of the Avanti J-30I centrifuge (instrument #s 363125, 363122, 363121, 363120, 363118), and compatible with rotors: SX4750 (part # 369702) SX4750A Aries (part # 369704).
Catalog No. NC0111689 Supplier Beckman Coulter Supplier No. 961648
Replacement O-ring for the centrifuge bucket cover assembly of the Avanti J-30I centrifuge (instrument #s 363125, 363122, 363121, 363120, 363118), and compatible with rotors: SX4750 (part # 369702) SX4750A Aries (part # 369704).