Learn More
Ace Glass, Inc. O-ring,Fetfe,size 112,pk/12
SDP
Catalog No. NC0139196
Encompass_Preferred
FETFE — A fluoroelastomer with special TFE Additives.Size I.D., W, Dia112mm 12.4mm 2.6mm
Catalog No. NC0139196 Supplier Ace Glass, Inc. Supplier No. 7855720
FETFE — A fluoroelastomer with special TFE Additives.Size I.D., W, Dia112mm 12.4mm 2.6mm