Learn More
Chemglass Life Sciences O-Ring, Viton, #113
SDP
Catalog No. 501531547
Encompass_Preferred
Support Tubes, Stainless Steel , 1/2in OD X 36in Length. These support tubes are made of brilliant polished 304 Stainless Steel . They have good surface hardness and do not score from clamp binding screws. Tubes are 1/2" (13mm) in diameter, and are hollow, reducing weight to 1/2 of solid Stainless Steel rods while maintaining all the advantages of the Stainless Steel . The walls are 1/16" (1.6mm) thick, strong, and do not sag. The ends of each tube are capped with polyethylene tight fitting end-caps.
Catalog No. 50-153-1547 Supplier Chemglass Life Sciences Supplier No. CG305113
Support Tubes, Stainless Steel , 1/2in OD X 36in Length. These support tubes are made of brilliant polished 304 Stainless Steel . They have good surface hardness and do not score from clamp binding screws. Tubes are 1/2" (13mm) in diameter, and are hollow, reducing weight to 1/2 of solid Stainless Steel rods while maintaining all the advantages of the Stainless Steel . The walls are 1/16" (1.6mm) thick, strong, and do not sag. The ends of each tube are capped with polyethylene tight fitting end-caps.