Learn More
Waters Corp Waters Corporation O-Ring, FKM, 2-030, MS System
Catalog No. 50792825 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement O-ring, model 2-030, is designed for use on the Source enclosure of 3100, SQD, and ACQUITY TQD mass spectrometry systems. It is made from FKM (Fluoroelastomer) for reliable performance.
  • Designed for mass spectrometry systems
  • Made from FKM material
  • Replacement part for 3100, SQD, and ACQUITY TQD systems
  • Supplied as one unit per package
Catalog No. 50-792-825 Supplier Waters Corp Supplier No. 700003066
This replacement O-ring, model 2-030, is designed for use on the Source enclosure of 3100, SQD, and ACQUITY TQD mass spectrometry systems. It is made from FKM (Fluoroelastomer) for reliable performance.
  • Designed for mass spectrometry systems
  • Made from FKM material
  • Replacement part for 3100, SQD, and ACQUITY TQD systems
  • Supplied as one unit per package