Learn More
Chemglass Life Sciences O-Ring, Viton, #219
SDP
Catalog No. 501531561
Encompass_Preferred
O-Ring, Viton, #219. Viton o-ring is stocked in brown 75 durometer. They are suitable for acids, aliphatic hydrocarbons, oxidizing agents and all petroleum products. Viton has a low compression set and high temperature resistance. Viton is especially good for vacuum service and low gas permeability and has a useful temperature range of -30 to 200 C. For complete chemical resistance and fluid compatibility, see chart in the specifications tab at the bottom of this listing. FOR VITON O-RINGS USED WITH DURAN FLANGES, SEE CG-147. 32.92 mm ID x 3.53mm Cross section.
Catalog No. 50-153-1561 Supplier Chemglass Life Sciences Supplier No. CG305219
O-Ring, Viton, #219. Viton o-ring is stocked in brown 75 durometer. They are suitable for acids, aliphatic hydrocarbons, oxidizing agents and all petroleum products. Viton has a low compression set and high temperature resistance. Viton is especially good for vacuum service and low gas permeability and has a useful temperature range of -30 to 200 C. For complete chemical resistance and fluid compatibility, see chart in the specifications tab at the bottom of this listing. FOR VITON O-RINGS USED WITH DURAN FLANGES, SEE CG-147. 32.92 mm ID x 3.53mm Cross section.