Learn More
Waters Corp Waters O-Ring, FKM, AS012, 3/pk
Supplier:  Waters Corp 5711915S
Encompass_Preferred
This replacement O-ring is typically used as a seal in legacy ESI and APcI probe designs. It is found in the probes of 3100, SQD, ACQUITY TQD, and earlier Mass Spectrometers, as well as earlier versions of Xevo and Synapt products before the introduction of Tool Free Probes.
  • Manufactured from FKM material.
  • Designed for use as a seal in legacy ESI and APcI probe designs.
  • Compatible with probes of 3100, SQD, ACQUITY TQD, and earlier Mass Spectrometers.
  • Suitable for earlier versions of Xevo and Synapt products.
  • Provided in a package of 3.
  • Replace as required.
Catalog No. 50-466-708