Learn More
Waters Corp Waters O-Ring, FKM, AS029, 5 pk
Supplier:  Waters Corp 5711036S
Encompass_Preferred
This O-ring serves as a replacement for the Ion Block Support. It is found in the source assemblies of SQD2, Xevo TQD, Xevo TQ-S, Xevo TQ-S Micro, Xevo TQ-S cronos, Xevo TQ-XS, Xevo TQ Absolute, Xevo G2-XS, Vion, SYNAPT G2-Si, SYNAPT XS, Select Series Cyclic IMS & MRT.
  • Made from FKM material
  • Designed for MS system types
  • Supplied in a 5 pack
  • Replacement is recommended as required
Catalog No. 50-466-707