Learn More
Waters Corp Waters Corporation O-Ring, FKM, AS035, MS
Catalog No. 50792470 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement O-ring is designed to AS035 specifications. It is typically found on Ion Block Assemblies for various mass spectrometry systems including SQD2, Xevo TQD, Xevo TQ-S, Xevo TQ-S Micro, Xevo TQ-S cronos, Xevo TQ-XS, Xevo TQ Absolute, Vion, SYNAPT XS, Select Series Cyclic IMS & MRT.
  • Designed as an O-ring.
  • Suitable for MS system type.
  • Constructed from FKM material.
  • Recommended for replacement as required.
  • Packaged with 1 unit per package.
Catalog No. 50-792-470 Supplier Waters Corp Supplier No. 700000904
This replacement O-ring is designed to AS035 specifications. It is typically found on Ion Block Assemblies for various mass spectrometry systems including SQD2, Xevo TQD, Xevo TQ-S, Xevo TQ-S Micro, Xevo TQ-S cronos, Xevo TQ-XS, Xevo TQ Absolute, Vion, SYNAPT XS, Select Series Cyclic IMS & MRT.
  • Designed as an O-ring.
  • Suitable for MS system type.
  • Constructed from FKM material.
  • Recommended for replacement as required.
  • Packaged with 1 unit per package.