Learn More
Waters Corp Waters O-Ring, FKM, BS006
Supplier:  Waters Corp 5711140
Encompass_Preferred
This replacement O-ring BS006 is designed for use primarily in the Reference Probe Tip Assembly on LockSpray Sources. Its FKM construction ensures compatibility with a range of mass spectrometers.
  • Designed for replacement O-ring BS006
  • Primarily found in the Reference Probe Tip Assembly on LockSpray Sources
  • Constructed from FKM material
  • Fits into Xevo G2-XS, Vion, Synapt XS, Select Series Cyclic IMS & MRT mass spectrometers
  • Also compatible with earlier Tof and Q-Tof instruments
  • System type: MS
  • Units per package: 1 pk
  • Replacement schedule: As required
Catalog No. 50-792-102