Learn More
Chemglass Life Sciences O-Ring, Viton, #009
SDP
Catalog No. 50873635
Encompass_Preferred
O-Ring, Viton, #009 12 PACK. Viton o-ring is stocked in brown 75 durometer. They are suitable for acids, aliphatic hydrocarbons, oxidizing agents and all petroleum products. Viton has a low compression set and high temperature resistance. Viton is especially good for vacuum service and low gas permeability and has a useful temperature range of -30 to 200¿ C. FOR VITON O-RINGS USED WITH DURAN FLANGES, SEE CG-147.
Catalog No. 50-873-635 Supplier Chemglass Life Sciences Supplier No. CG305009
O-Ring, Viton, #009 12 PACK. Viton o-ring is stocked in brown 75 durometer. They are suitable for acids, aliphatic hydrocarbons, oxidizing agents and all petroleum products. Viton has a low compression set and high temperature resistance. Viton is especially good for vacuum service and low gas permeability and has a useful temperature range of -30 to 200¿ C. FOR VITON O-RINGS USED WITH DURAN FLANGES, SEE CG-147.