Learn More
Chemglass Life Sciences Chemglass Viton O-Ring #010, Pack of 12 - Replacement seal for lab glassware joints and fittings
SDP
Catalog No. NC2290281
Encompass_Preferred
Chemglass Viton O-rings, size #010 (pack of 12), are chemical-resistant elastomer seals designed to provide durable, leak-resistant performance for laboratory glassware joints, fittings, and reaction vessels.
  • Viton material provides chemical resistance and durability.
  • Pack of 12 rings for maintenance and spare inventory.
  • Size #010 fits small laboratory joints and fittings.
  • Suitable for use with common laboratory solvents and reagents.
  • Designed for reliable, leak-resistant sealing under typical lab conditions.
This item is not returnable. View return policy
Catalog No. NC2290281 Supplier Chemglass Life Sciences Supplier No. CG305010PK
Chemglass Viton O-rings, size #010 (pack of 12), are chemical-resistant elastomer seals designed to provide durable, leak-resistant performance for laboratory glassware joints, fittings, and reaction vessels.
  • Viton material provides chemical resistance and durability.
  • Pack of 12 rings for maintenance and spare inventory.
  • Size #010 fits small laboratory joints and fittings.
  • Suitable for use with common laboratory solvents and reagents.
  • Designed for reliable, leak-resistant sealing under typical lab conditions.