Learn More
Chemglass Life Sciences Replacement Viton O-Rings for 11/25 PTFE Plug
SDP
Catalog No. CG45701
Encompass_Preferred
Replacement Viton O-Rings for 11/25 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.
Catalog No. CG45701 Supplier Chemglass Life Sciences Supplier No. CG45701
Replacement Viton O-Rings for 11/25 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.