Learn More
Chemglass Life Sciences Replacement Viton O-Rings for 15.2/30 PTFE Plug
SDP
Catalog No. CG45713
Encompass_Preferred
Replacement Viton O-Rings for 15.2/30 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.
Catalog No. CG45713 Supplier Chemglass Life Sciences Supplier No. CG45713
Replacement Viton O-Rings for 15.2/30 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.