Learn More
Ace Glass, Inc. O-RING VITON SIZE 007 O-RING VITON SIZE 007
SDP
Catalog No. 50979495
Encompass_Preferred
Size 007 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.
Catalog No. 50-979-495 Supplier Ace Glass, Inc. Supplier No. 785502
Size 007 Viton o-ring. Pack/12. The Viton class of elastomers is a family comprised of a linear co-polymer of vinylidine fluoride and hexafluoropropylene. Has generally good temperature and chemical resistance.